Product Details
- SNP ID
-
hCV25957603
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:58209160 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTGCTTTTGAAATAAAAAGAGAAG[C/T]AATAAAATTCTTACCAATTTTTAGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACTR10
PubMed Links
Gene Details
- Gene
- ACTR10
- Gene Name
- actin-related protein 10 homolog
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018477.2 |
|
Intron |
|
|
NP_060947.1 |
View Full Product Details