Product Details

SNP ID
rs34045351
Assay Type
Functionally Tested
NCBI dbSNP Submissions
11
Location
Chr.1:59815171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGCTGCCCCTGTGCGACAGCCT[C/T]ATGATCTGGGTGAGTGCGAGGAGGC
Phenotype
MIM: 607820
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HOOK1 PubMed Links

Gene Details

Gene
HOOK1
Gene Name
hook microtubule-tethering protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015888.4 250 Silent Mutation CTC,CTT L18L NP_056972.1
XM_006710676.1 250 Silent Mutation CTC,CTT L18L XP_006710739.1
XM_011541562.2 250 UTR 5 XP_011539864.1
XM_011541563.1 250 Silent Mutation CTC,CTT L18L XP_011539865.1
XM_017001424.1 250 Silent Mutation CTC,CTT L18L XP_016856913.1

View Full Product Details