Product Details

SNP ID
rs73204749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:19823211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCCCTATATATTTTAGATACTA[C/T]ATCAAATGAACTCTGAAAGCTTTTG
Phenotype
MIM: 611353
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INTS10 PubMed Links

Gene Details

Gene
INTS10
Gene Name
integrator complex subunit 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018142.2 Intron NP_060612.2
XM_005273555.3 Intron XP_005273612.1
XM_005273556.3 Intron XP_005273613.1
XM_005273557.3 Intron XP_005273614.1
XM_006716355.2 Intron XP_006716418.1
XM_017013606.1 Intron XP_016869095.1
XM_017013607.1 Intron XP_016869096.1
XM_017013608.1 Intron XP_016869097.1
XM_017013609.1 Intron XP_016869098.1

View Full Product Details