Product Details

SNP ID
rs142246587
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:112206716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCCCTTTTCTTTTCTGCCGGGT[A/G]ATGGCTGCTTCCAAGACCCAGGGGG
Phenotype
MIM: 607679 MIM: 605465
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DOCK4 PubMed Links

Gene Details

Gene
DOCK4
Gene Name
dedicator of cytokinesis 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014705.3 16 Intron NP_055520.3
XM_006716188.2 16 Intron XP_006716251.1
XM_006716189.2 16 Intron XP_006716252.1
XM_017012819.1 16 Intron XP_016868308.1
XM_017012820.1 16 Intron XP_016868309.1
XM_017012821.1 16 Intron XP_016868310.1
XM_017012822.1 16 Intron XP_016868311.1
XM_017012823.1 16 Intron XP_016868312.1
XM_017012824.1 16 Intron XP_016868313.1
XM_017012825.1 16 Intron XP_016868314.1
Gene
ZNF277
Gene Name
zinc finger protein 277
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021994.2 16 UTR 5 NP_068834.2
XM_005250129.3 16 UTR 5 XP_005250186.1
XM_011515768.2 16 UTR 5 XP_011514070.1
XM_017011720.1 16 UTR 5 XP_016867209.1

View Full Product Details