Product Details

SNP ID
rs34341878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133332256 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCAGCAGCTGCCGGTAGTTCCT[C/T]CCGGTCAGCGGCGTGAGGTTCCCCT
Phenotype
MIM: 185641 MIM: 185642
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED22 PubMed Links

Gene Details

Gene
MED22
Gene Name
mediator complex subunit 22
There are no transcripts associated with this gene.

Gene
SURF6
Gene Name
surfeit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278942.1 777 UTR 3 NP_001265871.1
NM_006753.5 777 Silent Mutation GGA,GGG G233G NP_006744.2

View Full Product Details