Product Details

SNP ID
rs8177368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126290571 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAACAGTTCCTCAGCTGGTCATG[C/T]TGAGCTCATACCCTGTAAGTCTGAC
Phenotype
MIM: 606252
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HP11014 PubMed Links

Gene Details

Gene
HP11014
Gene Name
uncharacterized HP11014
There are no transcripts associated with this gene.

Gene
TIRAP
Gene Name
TIR domain containing adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039661.1 367 UTR 5 NP_001034750.1
NM_001318776.1 367 UTR 5 NP_001305705.1
NM_001318777.1 367 UTR 5 NP_001305706.1
NM_148910.2 367 UTR 5 NP_683708.1
XM_005271399.3 367 Intron XP_005271456.2
XM_011542576.2 367 Intron XP_011540878.1
XM_017017161.1 367 Intron XP_016872650.1
XM_017017162.1 367 UTR 5 XP_016872651.1

View Full Product Details