Product Details

SNP ID
rs6702916
Assay Type
Functionally tested
NCBI dbSNP Submissions
48
Location
Chr.1:3816075 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGCCTTCGCACTGCACAGCCAGT[A/T]TGCAGGTGGAGAAGGATTTTGGTCA
Phenotype
MIM: 616690
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CEP104 PubMed Links

Gene Details

Gene
CEP104
Gene Name
centrosomal protein 104
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014704.3 Intron NP_055519.1
XM_005244815.4 Intron XP_005244872.1
XM_011542473.2 Intron XP_011540775.1
XM_011542474.2 Intron XP_011540776.1
XM_011542475.2 Intron XP_011540777.1
XM_011542476.2 Intron XP_011540778.1
XM_011542477.2 Intron XP_011540779.1
XM_011542478.1 Intron XP_011540780.1
XM_017002917.1 Intron XP_016858406.1
XM_017002918.1 Intron XP_016858407.1
XM_017002919.1 Intron XP_016858408.1

View Full Product Details