Product Details

SNP ID
rs149118751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81543996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTGGCCACCTTCCCCAGCGGGC[C/T]GACATACCTGCATGCGCCCGACAAC
Phenotype
MIM: 611301 MIM: 607643
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAAP100 PubMed Links

Gene Details

Gene
FAAP100
Gene Name
Fanconi anemia core complex associated protein 100
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025161.5 Intron NP_079437.5
XM_006722111.2 Intron XP_006722174.1
XM_006722112.3 Intron XP_006722175.1
XM_017025162.1 Intron XP_016880651.1
XM_017025163.1 Intron XP_016880652.1
Gene
FSCN2
Gene Name
fascin actin-bundling protein 2, retinal
There are no transcripts associated with this gene.

View Full Product Details