Product Details

SNP ID
rs74258162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35903343 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCCTCATCCACCTTCTTTCTCT[C/T]TCCACAGTGGCTGCAGCTCAGACGA
Phenotype
MIM: 604089 MIM: 604142
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCST PubMed Links

Gene Details

Gene
HCST
Gene Name
hematopoietic cell signal transducer
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007469.1 Intron NP_001007470.1
NM_014266.3 Intron NP_055081.1
XM_017026193.1 Intron XP_016881682.1
Gene
NFKBID
Gene Name
NFKB inhibitor delta
There are no transcripts associated with this gene.

Gene
TYROBP
Gene Name
TYRO protein tyrosine kinase binding protein
There are no transcripts associated with this gene.

View Full Product Details