Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_022092.2 | 270 | Intron | NP_071375.1 | ||
XM_005255470.1 | 270 | Intron | XP_005255527.1 | ||
XM_005255471.3 | 270 | Intron | XP_005255528.1 | ||
XM_011522572.1 | 270 | Intron | XP_011520874.1 | ||
XM_011522573.1 | 270 | Intron | XP_011520875.1 | ||
XM_017023532.1 | 270 | Intron | XP_016879021.1 | ||
XM_017023533.1 | 270 | Intron | XP_016879022.1 | ||
XM_017023534.1 | 270 | Intron | XP_016879023.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_016541.2 | 270 | Silent Mutation | AAC,AAT | N56N | NP_057625.1 |