Product Details

SNP ID
rs14532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:134157303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGCACTATTCTAGGTGCAATAAA[A/G]GGGAATCTTAACCTTAGAAATATGA
Phenotype
MIM: 600524
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RYK PubMed Links

Gene Details

Gene
RYK
Gene Name
receptor-like tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005861.2 2270 Intron NP_001005861.1
NM_002958.3 2270 Intron NP_002949.2
XM_017007016.1 2270 UTR 3 XP_016862505.1

View Full Product Details