Product Details
- SNP ID
-
rs7977270
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:65767615 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAAAAAAAAAAAAAAAAAATAGCA[C/T]ATCACCAAGAAAGAGCAAACAAACA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RPSAP52
PubMed Links
Gene Details
- Gene
- RPSAP52
- Gene Name
- ribosomal protein SA pseudogene 52
There are no transcripts associated with this gene.
View Full Product Details