Product Details

SNP ID
rs4237854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48930396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATGGGAGAGAGAAGAGTAAGGG[C/G]AATGTCCCACATGTTGCTCTGTATA
Phenotype
MIM: 103190 MIM: 611088 MIM: 610571
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARF3 PubMed Links

Gene Details

Gene
ARF3
Gene Name
ADP ribosylation factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001659.2 1627 Intron NP_001650.1
XM_005268856.1 1627 Intron XP_005268913.1
XM_006719391.3 1627 UTR 3 XP_006719454.1
Gene
CCDC65
Gene Name
coiled-coil domain containing 65
There are no transcripts associated with this gene.

Gene
FKBP11
Gene Name
FK506 binding protein 11
There are no transcripts associated with this gene.

View Full Product Details