Product Details

SNP ID
rs4409578
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:139739160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGAACTCTACCTCACTCAAATT[C/G]TGAAGAACACCTTGAATAATGTTCT
Phenotype
MIM: 300516
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATP11C PubMed Links

Gene Details

Gene
ATP11C
Gene Name
ATPase phospholipid transporting 11C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010986.2 Intron NP_001010986.1
NM_173694.4 Intron NP_775965.2

View Full Product Details