Product Details
- SNP ID
-
rs6689324
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
48
- Location
-
Chr.1:201985569 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACATGCCTGGCTTAATTGAGCAGC[C/T]AGTTCAGAGAGTTCCCATATACAGT
- Phenotype
-
MIM: 602675
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RNPEP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117262471] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RNPEP
- Gene Name
- arginyl aminopeptidase
View Full Product Details