Product Details

SNP ID
rs4394416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:52625902 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTAAAGCCTTTGAAAGAAGGCATA[C/T]GGATTTTTATTCCATACAAATAATT
Phenotype
MIM: 606837
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RB1CC1 PubMed Links

Gene Details

Gene
RB1CC1
Gene Name
RB1 inducible coiled-coil 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083617.1 Intron NP_001077086.1
NM_014781.4 Intron NP_055596.3
XM_011517643.2 Intron XP_011515945.1
XM_011517644.2 Intron XP_011515946.1
XM_011517645.1 Intron XP_011515947.1
XM_011517646.2 Intron XP_011515948.1
XM_011517647.2 Intron XP_011515949.1
XM_011517649.2 Intron XP_011515951.1
XM_017014103.1 Intron XP_016869592.1
XM_017014104.1 Intron XP_016869593.1
XM_017014105.1 Intron XP_016869594.1
XM_017014106.1 Intron XP_016869595.1
XM_017014107.1 Intron XP_016869596.1
XM_017014108.1 Intron XP_016869597.1
XM_017014109.1 Intron XP_016869598.1
XM_017014110.1 Intron XP_016869599.1
XM_017014111.1 Intron XP_016869600.1
XM_017014112.1 Intron XP_016869601.1

View Full Product Details