Product Details

SNP ID
rs11229411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58402869 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGAGCAAGAGAGAACCATGACTG[C/T]TGGAATATCACAGAAAAAGTGATGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5B3 PubMed Links

Gene Details

Gene
OR5B3
Gene Name
olfactory receptor family 5 subfamily B member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005469.1 541 Missense Mutation ACA,GCA T181A NP_001005469.1

View Full Product Details