Product Details

SNP ID
rs2914672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:90480226 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGGTGTATTGAAAAAGGAATGAG[A/G]CAGTGGTTAACTCAACCACAGTCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MBLAC2 PubMed Links

Gene Details

Gene
MBLAC2
Gene Name
metallo-beta-lactamase domain containing 2
There are no transcripts associated with this gene.

Gene
POLR3G
Gene Name
RNA polymerase III subunit G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006467.2 Intron NP_006458.2
XM_005248404.3 Intron XP_005248461.1
XM_011543100.2 Intron XP_011541402.1
XM_011543101.2 Intron XP_011541403.1
XM_011543102.2 Intron XP_011541404.1
XM_011543103.2 Intron XP_011541405.1
XM_011543104.2 Intron XP_011541406.1
XM_011543105.2 Intron XP_011541407.1
XM_017008957.1 Intron XP_016864446.1

View Full Product Details