Product Details

SNP ID
rs2272452
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:93045054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACGAGGGCGCCCAGGAGGGGCC[G/A]GGGGGCCAGGGGCAGCCCCGCAGCC
Phenotype
MIM: 607362
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
RGMA PubMed Links

Gene Details

Gene
RGMA
Gene Name
repulsive guidance molecule family member a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166283.1 1552 Silent Mutation NP_001159755.1
NM_001166286.1 1552 Silent Mutation NP_001159758.1
NM_001166287.1 1552 Silent Mutation NP_001159759.1
NM_001166288.1 1552 Silent Mutation NP_001159760.1
NM_001166289.1 1552 Silent Mutation NP_001159761.1
NM_020211.2 1552 Silent Mutation NP_064596.2

View Full Product Details