Product Details

SNP ID
rs6010225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50276112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCACTTGGGGCCCCCCATGTGGC[A/G]CAGCCTCAGACAGACACCCACGCAG
Phenotype
MIM: 602898 MIM: 604293
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAPK11 PubMed Links

Gene Details

Gene
MAPK11
Gene Name
mitogen-activated protein kinase 11
There are no transcripts associated with this gene.

Gene
PLXNB2
Gene Name
plexin B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012401.3 Intron NP_036533.2
XM_005261909.1 Intron XP_005261966.1
XM_005261910.2 Intron XP_005261967.1
XM_005261911.4 Intron XP_005261968.1
XM_006724413.2 Intron XP_006724476.1
XM_011530682.2 Intron XP_011528984.1
XM_017028703.1 Intron XP_016884192.1
XM_017028704.1 Intron XP_016884193.1

View Full Product Details