Product Details

SNP ID
rs2447196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43601620 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGAGTCATACTGCTGGGTTTTAA[A/G]TCTTGCCTCTGCCCTTAGCTGTATA
Phenotype
MIM: 123290 MIM: 606440
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CKMT1B PubMed Links

Gene Details

Gene
CKMT1B
Gene Name
creatine kinase, mitochondrial 1B
There are no transcripts associated with this gene.

Gene
STRC
Gene Name
stereocilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153700.2 Intron NP_714544.1
XM_011521278.2 Intron XP_011519580.1
XM_017021954.1 Intron XP_016877443.1
XM_017021955.1 Intron XP_016877444.1
XM_017021956.1 Intron XP_016877445.1
XM_017021957.1 Intron XP_016877446.1
XM_017021958.1 Intron XP_016877447.1
XM_017021959.1 Intron XP_016877448.1
XM_017021960.1 Intron XP_016877449.1
XM_017021961.1 Intron XP_016877450.1
XM_017021962.1 Intron XP_016877451.1

View Full Product Details