Product Details

SNP ID
rs4075586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:91855591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTGTGTCTATAGTTATAATATG[C/T]AGTTGCAGACAGTACCTATTAGCCC
Phenotype
MIM: 612435
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLCO3A1 PubMed Links

Gene Details

Gene
SLCO3A1
Gene Name
solute carrier organic anion transporter family member 3A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145044.1 Intron NP_001138516.1
NM_013272.3 Intron NP_037404.2
XM_005254889.1 Intron XP_005254946.1
XM_005254891.2 Intron XP_005254948.1
XM_011521456.2 Intron XP_011519758.1

View Full Product Details