Product Details

SNP ID
rs1112462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89204321 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATCTGGTGTTACGTAAGAGTACA[C/G]CTGCATTCTTTTGCATGAAGATATT
Phenotype
MIM: 114019
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDH15 PubMed Links

Gene Details

Gene
CDH15
Gene Name
cadherin 15
There are no transcripts associated with this gene.

Gene
SLC22A31
Gene Name
solute carrier family 22 member 31
There are no transcripts associated with this gene.

Gene
ZNF778
Gene Name
zinc finger protein 778
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201407.1 Intron NP_001188336.1
NM_182531.3 Intron NP_872337.2
XM_005256288.2 Intron XP_005256345.1
XM_011522940.2 Intron XP_011521242.1
XM_017023015.1 Intron XP_016878504.1
XM_017023016.1 Intron XP_016878505.1
XM_017023017.1 Intron XP_016878506.1

View Full Product Details