Product Details

SNP ID
rs2137841
Assay Type
DME
NCBI dbSNP Submissions
19
Location
Chr.1:225844618 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
GACAGGGCTGGATGACCCAGAAGCA[A/T]GAGCGGTGAGCCTGGCTGAGCCGAG
Phenotype
MIM: 132810
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
EPHX1 PubMed Links

Gene Details

Gene
EPHX1
Gene Name
epoxide hydrolase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000120.3 1410 Missense Mutation CAA,CAT Q387H NP_000111.1
NM_001136018.3 1410 Missense Mutation CAA,CAT Q387H NP_001129490.1
NM_001291163.1 1410 Missense Mutation CAA,CAT Q387H NP_001278092.1
Gene
TMEM63A
Gene Name
transmembrane protein 63A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014698.2 1410 Intron NP_055513.2
XM_006711841.3 1410 Intron XP_006711904.1
XM_011544328.2 1410 Intron XP_011542630.1
XM_011544329.2 1410 Intron XP_011542631.1
XM_011544330.2 1410 Intron XP_011542632.1
XM_011544331.2 1410 Intron XP_011542633.1
XM_011544332.2 1410 Intron XP_011542634.1
XM_017002916.1 1410 Intron XP_016858405.1

View Full Product Details