Product Details

SNP ID
rs6672410
Assay Type
Functionally tested
NCBI dbSNP Submissions
39
Location
Chr.1:234632166 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAGGATAAATTCTCTTCTGGGGT[G/T]GTTAAACCAATAGAAAATTAGCCTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00184 PubMed Links

Gene Details

Gene
LINC00184
Gene Name
long intergenic non-protein coding RNA 184
There are no transcripts associated with this gene.

View Full Product Details