Product Details

SNP ID
rs11993916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144280341 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGAGAATCACTGGGCAGCCCCTTT[C/T]CAGACTCCACAGAGCTGCAGCTCCA
Phenotype
MIM: 610596
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BOP1 PubMed Links

Gene Details

Gene
BOP1
Gene Name
block of proliferation 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015201.4 Intron NP_056016.1

View Full Product Details