Product Details

SNP ID
rs2250064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102750548 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTCTTTTTTTTTTTCTTTTGAAA[C/T]GGTGTCTTGCTCCGTCGCCCAGGTT
Phenotype
MIM: 611129
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WBP1L PubMed Links

Gene Details

Gene
WBP1L
Gene Name
WW domain binding protein 1-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083913.1 Intron NP_001077382.1
NM_017787.4 Intron NP_060257.4
XM_011539913.2 Intron XP_011538215.1
XM_017016360.1 Intron XP_016871849.1

View Full Product Details