Product Details

SNP ID
rs4090240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:74504071 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTTCAAACCACTTAGAATTTTGG[C/T]TTGCTCAGGTAACCTCTAAAATATA
Phenotype
MIM: 601972
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RORB PubMed Links

Gene Details

Gene
RORB
Gene Name
RAR related orphan receptor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006914.3 Intron NP_008845.2
Gene
RORB-AS1
Gene Name
RORB antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details