Product Details

SNP ID
rs4098047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:74509768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAAAGGCCTATTATCATCCTTCT[A/G]TGTTGTGTACAAGCATAAGATGTTT
Phenotype
MIM: 601972
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RORB PubMed Links

Gene Details

Gene
RORB
Gene Name
RAR related orphan receptor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006914.3 Intron NP_008845.2

View Full Product Details