Product Details

SNP ID
rs7322498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:108220173 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACCTATCTCTTAACAGACCATGAC[C/T]CAAAACACTGTCTCCTTTCCATTGC
Phenotype
MIM: 601837
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD13 PubMed Links

Gene Details

Gene
ABHD13
Gene Name
abhydrolase domain containing 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032859.2 Intron NP_116248.2
XM_011521128.2 Intron XP_011519430.1
Gene
LIG4
Gene Name
DNA ligase 4
There are no transcripts associated with this gene.

View Full Product Details