Product Details

SNP ID
rs4583952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:83072527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCATCTTGGATGCATAGCATTTA[A/T]TTCACAAACAAGTGGTTTCTTTCCT
Phenotype
MIM: 616823 MIM: 612495
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DOPEY1 PubMed Links

Gene Details

Gene
DOPEY1
Gene Name
dopey family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199942.1 Intron NP_001186871.1
NM_015018.3 Intron NP_055833.2
XM_011535619.2 Intron XP_011533921.1
XM_017010559.1 Intron XP_016866048.1
XM_017010560.1 Intron XP_016866049.1
XM_017010561.1 Intron XP_016866050.1
XM_017010562.1 Intron XP_016866051.1
XM_017010563.1 Intron XP_016866052.1
XM_017010564.1 Intron XP_016866053.1
XM_017010565.1 Intron XP_016866054.1
XM_017010566.1 Intron XP_016866055.1
XM_017010567.1 Intron XP_016866056.1
XM_017010568.1 Intron XP_016866057.1
XM_017010569.1 Intron XP_016866058.1
XM_017010570.1 Intron XP_016866059.1
XM_017010571.1 Intron XP_016866060.1
XM_017010572.1 Intron XP_016866061.1
XM_017010573.1 Intron XP_016866062.1
XM_017010574.1 Intron XP_016866063.1
Gene
UBE3D
Gene Name
ubiquitin protein ligase E3D
There are no transcripts associated with this gene.

View Full Product Details