Product Details
- SNP ID
-
rs3741277
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:6681178 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGACTGATGGCTCCTTTCTAAGTC[C/T]TTCCATAACTACTACCCTACTAAAG
- Phenotype
-
MIM: 611830
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MRPL17
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144975670] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MRPL17
- Gene Name
- mitochondrial ribosomal protein L17
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022061.3 |
1573 |
UTR 3 |
|
|
NP_071344.1 |
View Full Product Details