Product Details

SNP ID
rs3751928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:73285213 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCACGCAGCCTAAGTGCAGGGAGC[A/G]TGATGAAGTCAGGCAGCCAGTCGGC
Phenotype
MIM: 605468
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC42EP4 PubMed Links

Gene Details

Gene
CDC42EP4
Gene Name
CDC42 effector protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012121.4 1827 UTR 3 NP_036253.2
XM_005257182.2 1827 UTR 3 XP_005257239.1

View Full Product Details