Product Details

SNP ID
rs3767734
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:198646877 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
AATCTTCTCAATGTAAAATAGTGTG[C/T]GGGGAAAGTGTCAAATCATCCTACT
Phenotype
MIM: 151460
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTPRC PubMed Links

Gene Details

Gene
PTPRC
Gene Name
protein tyrosine phosphatase, receptor type C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267798.1 Intron NP_001254727.1
NM_002838.4 Intron NP_002829.3
NM_080921.3 Intron NP_563578.2
XM_006711472.3 Intron XP_006711535.1
XM_006711473.3 Intron XP_006711536.1
XM_006711474.3 Intron XP_006711537.1

View Full Product Details