Product Details

SNP ID
rs3743869
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2163256 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGGAGTGTGGGTGACCTGCAC[A/G]GGTTCCAGGGGCGGGCTCATCTCTC
Phenotype
MIM: 605455 MIM: 606692
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
RAB26 PubMed Links
Additional Information
For this assay, SNP(s) [rs111638155] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
RAB26
Gene Name
RAB26, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
SNHG19
Gene Name
small nucleolar RNA host gene 19
There are no transcripts associated with this gene.

Gene
SNORD60
Gene Name
small nucleolar RNA, C/D box 60
There are no transcripts associated with this gene.

Gene
TRAF7
Gene Name
TNF receptor associated factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032271.2 Intron NP_115647.2
XM_005255627.4 Intron XP_005255684.1
XM_011522700.1 Intron XP_011521002.1

View Full Product Details