Product Details
- SNP ID
-
rs3815138
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:47784682 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTGGGAGGTGGGACGAACACAACA[C/T]GGGAGAACTGACCATCACCACGGCC
- Phenotype
-
MIM: 606542
MIM: 612187
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HDAC7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144355039] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HDAC7
- Gene Name
- histone deacetylase 7
- Gene
- SLC48A1
- Gene Name
- solute carrier family 48 member 1
There are no transcripts associated with this gene.
View Full Product Details