Product Details

SNP ID
rs3819097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66338389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGGCTGTTGCCTCAGTGCTGTG[A/C]CCCCAGGACCGGGAGCATAGGCCCC
Phenotype
MIM: 605517 MIM: 606259 MIM: 605965
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
B4GAT1 PubMed Links

Gene Details

Gene
B4GAT1
Gene Name
beta-1,4-glucuronyltransferase 1
There are no transcripts associated with this gene.

Gene
BRMS1
Gene Name
breast cancer metastasis suppressor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024957.1 Intron NP_001020128.1
NM_015399.3 Intron NP_056214.1
Gene
LOC102724064
Gene Name
uncharacterized LOC102724064
There are no transcripts associated with this gene.

Gene
RIN1
Gene Name
Ras and Rab interactor 1
There are no transcripts associated with this gene.

View Full Product Details