Product Details

SNP ID
rs3824070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:1535741 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTCCTGAAAGCACAGTTCTGGACG[A/G]CTGACCCTGGGCTGCCTGCCCTGCC
Phenotype
MIM: 600197
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100128653 PubMed Links

Gene Details

Gene
LOC100128653
Gene Name
uncharacterized LOC100128653
There are no transcripts associated with this gene.

Gene
MAFK
Gene Name
MAF bZIP transcription factor K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002360.3 Intron NP_002351.1
XM_005249851.2 Intron XP_005249908.2
XM_006715773.2 Intron XP_006715836.1
Gene
TMEM184A
Gene Name
transmembrane protein 184A
There are no transcripts associated with this gene.

View Full Product Details