Product Details

SNP ID
rs3930740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8391641 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATTGAAAGGTGTTGGTTGACTTA[C/T]GGACCTCAATAAACAGCCAATAGCT
Phenotype
MIM: 603704
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF222 PubMed Links

Gene Details

Gene
RNF222
Gene Name
ring finger protein 222
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146684.2 2130 UTR 3 NP_001140156.1
XM_011523978.2 2130 Intron XP_011522280.1
XM_011523979.2 2130 Intron XP_011522281.1
XM_011523980.2 2130 Intron XP_011522282.1
XM_011523981.2 2130 Intron XP_011522283.1
Gene
RPL26
Gene Name
ribosomal protein L26
There are no transcripts associated with this gene.

View Full Product Details