Product Details

SNP ID
rs3812399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143062710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGAACCACCACTGTGAGGTGA[C/G]AGGCACAGGTGGAGAAGGCCTTCTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR6V1 PubMed Links

Gene Details

Gene
OR6V1
Gene Name
olfactory receptor family 6 subfamily V member 1
There are no transcripts associated with this gene.

Gene
OR6W1P
Gene Name
olfactory receptor family 6 subfamily W member 1 pseudogene
There are no transcripts associated with this gene.

View Full Product Details