Product Details
- SNP ID
-
rs3736923
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:103090812 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTAAAACAAAGAACAAGATTGATT[C/T]TTGGCTCATTCAACAAATATTTATT
- Phenotype
-
MIM: 607803
MIM: 600417
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CNNM2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3736924] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CNNM2
- Gene Name
- cyclin and CBS domain divalent metal cation transport mediator 2
There are no transcripts associated with this gene.
- Gene
- NT5C2
- Gene Name
- 5'-nucleotidase, cytosolic II
View Full Product Details