Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_002943.3 | 9710 | UTR 3 | NP_002934.1 | ||
NM_134260.2 | 9710 | UTR 3 | NP_599022.1 | ||
NM_134261.2 | 9710 | UTR 3 | NP_599023.1 | ||
NM_134262.2 | 9710 | UTR 3 | NP_599024.1 | ||
XM_005254584.4 | 9710 | UTR 3 | XP_005254641.1 | ||
XM_011521874.1 | 9710 | UTR 3 | XP_011520176.1 | ||
XM_011521875.1 | 9710 | UTR 3 | XP_011520177.1 | ||
XM_011521877.2 | 9710 | UTR 3 | XP_011520179.1 | ||
XM_011521878.1 | 9710 | UTR 3 | XP_011520180.1 | ||
XM_011521879.2 | 9710 | UTR 3 | XP_011520181.1 | ||
XM_017022466.1 | 9710 | UTR 3 | XP_016877955.1 | ||
XM_017022467.1 | 9710 | UTR 3 | XP_016877956.1 |