Product Details

SNP ID
rs3759878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:44736389 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGCTATGAACCTTAAGATTAGAC[C/T]ACTAACTCGAATCTAAATGAGCTGC
Phenotype
MIM: 616017
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100419583 PubMed Links

Gene Details

Gene
LOC100419583
Gene Name
ring finger protein 4 pseudogene
There are no transcripts associated with this gene.

Gene
TRIM69
Gene Name
tripartite motif containing 69
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301144.1 28 UTR 5 NP_001288073.1
NM_001301145.1 28 UTR 5 NP_001288074.1
NM_001301146.1 28 UTR 5 NP_001288075.1
NM_080745.4 28 UTR 5 NP_542783.2
NM_182985.4 28 UTR 5 NP_892030.3

View Full Product Details