Product Details

SNP ID
rs115912907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:14632873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCCACTCGCCACCGCCGCTTCC[G/T]CAGCACCCATGGGGACCAGGAGACT
Phenotype
MIM: 611234
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM84A PubMed Links

Gene Details

Gene
FAM84A
Gene Name
family with sequence similarity 84 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145175.2 188 UTR 5 NP_660158.2
Gene
LOC653602
Gene Name
uncharacterized LOC653602
There are no transcripts associated with this gene.

View Full Product Details