Product Details

SNP ID
hCV27830355
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75064915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGAGGTTATGTCTGCTTCTGGG[C/T]TTCTGGAAGCTACTCACCGGTCTCC
Phenotype
MIM: 600875
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACYP1 PubMed Links

Gene Details

Gene
ACYP1
Gene Name
acylphosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001107.4 Intron NP_001098.1
NM_001302616.1 Intron NP_001289545.1
NM_001302617.1 Intron NP_001289546.1
Gene
ZC2HC1C
Gene Name
zinc finger C2HC-type containing 1C
There are no transcripts associated with this gene.

View Full Product Details