Product Details

SNP ID
rs76227487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:108569856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGCAGAAGCCAATGGGGAAGC[C/G]TTTCGTGTAGGTCTTCTGAGGTGGT
Phenotype
MIM: 602634 MIM: 612534
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAJB9 PubMed Links

Gene Details

Gene
DNAJB9
Gene Name
DnaJ heat shock protein family (Hsp40) member B9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012328.2 112 UTR 5 NP_036460.1
Gene
THAP5
Gene Name
THAP domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130475.2 112 Intron NP_001123947.1
NM_001287598.1 112 Intron NP_001274527.1
NM_001287599.1 112 Intron NP_001274528.1
NM_001287601.1 112 Intron NP_001274530.1
NM_182529.3 112 Intron NP_872335.2

View Full Product Details