Product Details

SNP ID
rs9882171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:40477330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGCCCTCGACCTTCCTGCTCCGA[A/G]GAGCTGGCCTGATGTCCTCGGGTCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF619 PubMed Links

Gene Details

Gene
ZNF619
Gene Name
zinc finger protein 619
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145082.2 199 UTR 5 NP_001138554.1
NM_001145083.1 199 UTR 5 NP_001138555.1
NM_001145093.2 199 UTR 5 NP_001138565.1
XM_006713112.2 199 UTR 5 XP_006713175.1
XM_006713113.3 199 UTR 5 XP_006713176.1
XM_011533607.1 199 UTR 5 XP_011531909.1
XM_011533608.1 199 UTR 5 XP_011531910.1
XM_017006225.1 199 UTR 5 XP_016861714.1

View Full Product Details