Product Details

SNP ID
rs77141819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44719855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTAACTGGCTTTTCATATAGAT[G/T]ATTGTATGATGGCCAGAGTGTACTA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF501 PubMed Links

Gene Details

Gene
ZNF501
Gene Name
zinc finger protein 501
There are no transcripts associated with this gene.

Gene
ZNF502
Gene Name
zinc finger protein 502
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134440.1 1091 Intron NP_001127912.1
NM_001134441.1 1091 Intron NP_001127913.1
NM_001134442.1 1091 Intron NP_001127914.1
NM_001282880.1 1091 Intron NP_001269809.1
NM_033210.4 1091 Intron NP_149987.2
XM_017007440.1 1091 UTR 5 XP_016862929.1

View Full Product Details