Product Details
- SNP ID
-
hCV27847037
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:9471110 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGACAAATTGTGCCCATCAGTTT[C/G]AATTGTTGAATTATAAGAAGCATGC
- Phenotype
-
MIM: 603433
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF143
PubMed Links
Gene Details
- Gene
- ZNF143
- Gene Name
- zinc finger protein 143
View Full Product Details