Product Details

SNP ID
rs114618346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43158582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGCTCTCTGTCTCCGGTGCATG[A/G]GGAAGGGGCGGTGGTTTTACTGGGG
Phenotype
MIM: 109610
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TSPO PubMed Links

Gene Details

Gene
TSPO
Gene Name
translocator protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000714.5 Intron NP_000705.2
NM_001256530.1 Intron NP_001243459.1
NM_001256531.1 Intron NP_001243460.1
Gene
TTLL12
Gene Name
tubulin tyrosine ligase like 12
There are no transcripts associated with this gene.

View Full Product Details